siRNA Details
Gene Symbol: KIAA1257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020741.2 | NP_065792.1 | 7 | 1179 | ||
XM_011513050.2 | XP_011511352.2 | 7 | 1079 | |||
XM_017006939.1 | XP_016862428.1 | 8 | 1634 | |||
XM_017006940.1 | XP_016862429.1 | 8 | 1630 | |||
XM_017006941.1 | XP_016862430.1 | 8 | 1207 | |||
XM_017006942.1 | XP_016862431.1 | 8 | 1601 | |||
XM_017006943.1 | XP_016862432.1 | 8 | 1359 | |||
XM_017006944.1 | XP_016862433.1 | 7 | 1179 | |||
XM_017006945.1 | XP_016862434.1 | 7 | 1079 | |||
XM_017006946.1 | XP_016862435.1 | 6 | 790 | |||
XM_017006947.1 | XP_016862436.1 | 5 | 674 | |||
XM_017006948.1 | XP_016862437.1 | 7 | 851 | |||
XM_017006949.1 | XP_016862438.1 | 6 | 798 | |||
XM_017006950.1 | XP_016862439.1 | 6 | 6285 | |||
XM_017006951.1 | XP_016862440.1 | 6 | 6390 | |||
XM_017006952.1 | XP_016862441.1 | 5 | 568 | |||
XM_017006953.1 | XP_016862442.1 | 5 | 574 | |||
GenBank mRNA | AB033083.1 | - | 7 | 1151 | ||
BC031632.1 | - | 6 | 1140 | 5170293 | ||
BC038736.1 | - | 7 | 1112 |
Gene Symbol: LOC100132731 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056377.1 | - | 6 | 1755 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
