siRNA Details
Gene Symbol: KIAA1257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020741.2 | NP_065792.1 | 8 | 1440 | ||
GenBank mRNA | AB033083.1 | - | 8 | 1412 | ||
BC031632.1 | - | 7 | 1401 | 5170293 | ||
BC038736.1 | - | 8 | 1373 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
