siRNA Details
Gene Symbol: ZFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016107.3 | NP_057191.2 | 20 | 3745 | ||
GenBank mRNA | AF100742.1 | - | 10 | 1740 | ||
AK025481.1 | - | 8 | 1438 | |||
AK055938.1 | - | 3 | 1494 | |||
AK123306.1 | - | 4 | 2142 | |||
AK307808.1 | - | 6 | 1084 | |||
AL137258.1 | - | 3 | 760 | |||
BC051893.1 | - | 13 | 2335 | |||
BC062986.1 | - | 13 | 2260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
