siRNA Details
Gene Symbol: TENM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080477.3 | NP_001073946.1 | 18, 19 | 3498 | ||
XM_017008385.1 | XP_016863874.1 | 22, 23 | 3963 | |||
XM_017008386.1 | XP_016863875.1 | 19, 20 | 3550 | |||
XM_017008387.1 | XP_016863876.1 | 19, 20 | 3523 | |||
XM_017008388.1 | XP_016863877.1 | 19, 20 | 3498 | |||
XM_017008389.1 | XP_016863878.1 | 22, 23 | 3963 | |||
XM_017008390.1 | XP_016863879.1 | 22, 23 | 3963 | |||
XM_017008391.1 | XP_016863880.1 | 22, 23 | 3963 | |||
XM_017008392.1 | XP_016863881.1 | 21, 22 | 3936 | |||
XM_017008393.1 | XP_016863882.1 | 21, 22 | 3936 | |||
XM_017008394.1 | XP_016863883.1 | 21, 22 | 3684 | |||
XM_017008395.1 | XP_016863884.1 | 17, 18 | 2936 | |||
XM_017008396.1 | XP_016863885.1 | 17, 18 | 2971 | |||
GenBank mRNA | AB040888.2 | - | 15, 16 | 2647 | ||
AK125869.1 | - | 10, 11 | 1764 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
