siRNA Details
Gene Symbol: SLF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032290.3 | NP_115666.2 | 4 | 288 | ||
XM_017009978.1 | XP_016865467.1 | 4 | 469 | |||
XM_017009979.1 | XP_016865468.1 | 4 | 5050 | |||
XM_017009980.1 | XP_016865469.1 | 4 | 470 | |||
XM_017009981.1 | XP_016865470.1 | 4 | 320 | |||
XM_017009982.1 | XP_016865471.1 | 4 | 469 | |||
GenBank mRNA | AK297452.1 | - | 4 | 296 | ||
AK299080.1 | - | 4 | 314 | |||
AL050298.1 | - | 4 | 293 | |||
BC054885.1 | - | 4 | 290 | |||
BC070332.1 | - | 4 | 326 | |||
BC107571.1 | - | 4 | 288 | |||
CR936768.1 | - | 4 | 294 | |||
DR001089.1 | - | 4 | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
