siRNA Details
Gene Symbol: TET2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127208.2 | NP_001120680.1 | 10 | 4696 | ||
XM_005263082.2 | XP_005263139.1 | 9 | 4572 | |||
XM_006714242.3 | XP_006714305.1 | 8 | 4240 | |||
GenBank mRNA | AB046766.1 | - | 3 | 256 | ||
AB075496.1 | - | 7 | 1750 | |||
BC110509.1 | - | 5 | 620 | |||
BC110510.2 | - | 5 | 620 | |||
FM992369.1 | - | 10 | 5069 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
