siRNA Details
Gene Symbol: RBM27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018989.1 | NP_061862.1 | 10, 11 | 1765 | ||
XM_005268466.1 | XP_005268523.1 | 10, 11 | 1818 | |||
XM_005268467.1 | XP_005268524.1 | 9, 10 | 1653 | |||
XM_005268468.1 | XP_005268525.1 | 9, 10 | 1650 | |||
XM_006714787.1 | XP_006714850.1 | 11, 12 | 1782 | |||
XM_011537661.1 | XP_011535963.1 | 10, 11 | 1818 | |||
XM_011537662.1 | XP_011535964.1 | 10, 11 | 1815 | |||
XM_017009587.1 | XP_016865076.1 | 11, 12 | 1779 | |||
GenBank mRNA | AB037732.1 | - | 6, 7 | 1088 | ||
AK128738.1 | - | 9, 10 | 1650 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
