siRNA Details
Gene Symbol: RBM27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018989.1 | NP_061862.1 | 4, 5 | 564 | ||
XM_005268466.1 | XP_005268523.1 | 4, 5 | 614 | |||
XM_005268467.1 | XP_005268524.1 | 4, 5 | 614 | |||
XM_005268468.1 | XP_005268525.1 | 4, 5 | 614 | |||
XM_006714787.1 | XP_006714850.1 | 4, 5 | 614 | |||
XM_011537661.1 | XP_011535963.1 | 4, 5 | 614 | |||
XM_011537662.1 | XP_011535964.1 | 4, 5 | 614 | |||
XM_017009587.1 | XP_016865076.1 | 4, 5 | 614 | |||
GenBank mRNA | AK128738.1 | - | 4, 5 | 614 | ||
BC009537.1 | - | 4, 5 | 567 | |||
BX647384.1 | - | 4, 5 | 564 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
