siRNA Details
Gene Symbol: AVL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015060.2 | NP_055875.1 | 10 | 1404 | ||
XM_005249668.2 | XP_005249725.1 | 10 | 1341 | |||
XM_005249669.2 | XP_005249726.1 | 10 | 1341 | |||
XM_005249670.2 | XP_005249727.1 | 10 | 1047 | |||
XM_005249671.4 | XP_005249728.1 | 10 | 1341 | |||
XM_011515227.2 | XP_011513529.1 | 10 | 1341 | |||
XM_017011891.1 | XP_016867380.1 | 10 | 1047 | |||
XM_017011892.1 | XP_016867381.1 | 10 | 1057 | |||
GenBank mRNA | AK090619.1 | - | 10 | 1266 | ||
BC027724.2 | - | 10 | 1256 | 4811237 | ||
D87682.1 | - | 8 | 913 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
