siRNA Details
Gene Symbol: AVL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015060.2 | NP_055875.1 | 3 | 574 | ||
XM_005249668.2 | XP_005249725.1 | 3 | 511 | |||
XM_005249669.2 | XP_005249726.1 | 3 | 511 | |||
XM_005249670.2 | XP_005249727.1 | 3 | 511 | |||
XM_005249671.4 | XP_005249728.1 | 3 | 511 | |||
XM_011515227.2 | XP_011513529.1 | 3 | 511 | |||
XM_017011891.1 | XP_016867380.1 | 3 | 511 | |||
XM_017011892.1 | XP_016867381.1 | 3 | 521 | |||
GenBank mRNA | AK090619.1 | - | 3 | 436 | ||
BC027724.2 | - | 3 | 426 | 4811237 | ||
D87682.1 | - | 1 | 83 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_030235, AK020003, BC117878, AK147365, AK122228, and others.
Gene Ontology Categories:
