siRNA Details
Gene Symbol: SEC24A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252231.1 | NP_001239160.1 | 2 | 466 | ||
NM_021982.2 | NP_068817.1 | 2 | 466 | |||
XM_006714523.2 | XP_006714586.1 | 2 | 841 | |||
XM_017008961.1 | XP_016864450.1 | 2 | 111 | |||
XM_017008963.1 | XP_016864452.1 | 2 | 841 | |||
GenBank mRNA | AJ131244.1 | - | 2 | 130 | ||
AK304060.1 | - | 2 | 466 | |||
BC019341.1 | - | 2 | 427 | 3355949 | ||
CD109783.1 | - | 2 | 489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
