siRNA Details
Gene Symbol: SEC24A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252231.1 | NP_001239160.1 | 5, 6 | 1280 | ||
NM_021982.2 | NP_068817.1 | 5, 6 | 1280 | |||
XM_006714523.2 | XP_006714586.1 | 4, 5 | 1481 | |||
XM_017008961.1 | XP_016864450.1 | 5, 6 | 925 | |||
XM_017008962.1 | XP_016864451.1 | 4, 5 | 585 | |||
XM_017008963.1 | XP_016864452.1 | 4, 5 | 1481 | |||
GenBank mRNA | AJ131244.1 | - | 5, 6 | 943 | ||
AK304060.1 | - | 5, 6 | 1269 | |||
BC019341.1 | - | 5, 6 | 1241 | 3355949 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
