siRNA Details
Gene Symbol: ZMIZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300959.1 | NP_001287888.1 | 18 | 3542 | ||
NM_031449.3 | NP_113637.3 | 19 | 3711 | |||
NM_174929.2 | NP_777589.2 | 17 | 3572 | |||
XM_005249866.2 | XP_005249923.1 | 19 | 3762 | |||
XM_005249867.4 | XP_005249924.1 | 19 | 3764 | |||
XM_005249868.4 | XP_005249925.1 | 19 | 4782 | |||
XM_005249869.2 | XP_005249926.1 | 19 | 3735 | |||
XM_005249870.2 | XP_005249927.1 | 18 | 3684 | |||
XM_005249871.2 | XP_005249928.1 | 19 | 3666 | |||
XM_005249872.2 | XP_005249929.1 | 18 | 3657 | |||
XM_005249873.2 | XP_005249930.1 | 19 | 3639 | |||
XM_011515566.1 | XP_011513868.1 | 13 | 2902 | |||
XM_017012674.1 | XP_016868163.1 | 19 | 3843 | |||
GenBank mRNA | AB067473.1 | - | 13 | 2795 | ||
AK057115.1 | - | 4 | 2172 | |||
AK057378.1 | - | 1 | 576 | |||
AK090415.1 | - | 19 | 3711 | |||
AK090841.1 | - | 11 | 2991 | |||
AK128359.1 | - | 1 | 1593 | |||
AL136572.1 | - | 9 | 2164 | |||
AL831933.1 | - | 10 | 3309 | |||
AY426594.1 | - | 17 | 3534 | |||
BC021924.1 | - | 13 | 2643 | |||
BC110435.1 | - | 18 | 3527 | 5518312 | ||
BC131780.1 | - | 4 | 1248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
