siRNA Details
Gene Symbol: ZMIZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300959.1 | NP_001287888.1 | 12 | 1804 | ||
NM_031449.3 | NP_113637.3 | 13 | 1973 | |||
NM_174929.2 | NP_777589.2 | 11 | 1834 | |||
XM_005249866.2 | XP_005249923.1 | 13 | 2024 | |||
XM_005249867.4 | XP_005249924.1 | 13 | 2026 | |||
XM_005249868.4 | XP_005249925.1 | 13 | 3044 | |||
XM_005249869.2 | XP_005249926.1 | 13 | 1997 | |||
XM_005249870.2 | XP_005249927.1 | 12 | 1946 | |||
XM_005249871.2 | XP_005249928.1 | 13 | 1928 | |||
XM_005249872.2 | XP_005249929.1 | 12 | 1919 | |||
XM_005249873.2 | XP_005249930.1 | 13 | 1901 | |||
XM_011515566.1 | XP_011513868.1 | 7 | 1164 | |||
XM_017012674.1 | XP_016868163.1 | 13 | 2105 | |||
GenBank mRNA | AB067473.1 | - | 7 | 1057 | ||
AF357908.1 | - | 12 | 1856 | |||
AK090415.1 | - | 13 | 1973 | |||
AK090841.1 | - | 5 | 1253 | |||
AL136572.1 | - | 3 | 426 | |||
AL831933.1 | - | 4 | 1571 | |||
AY426594.1 | - | 11 | 1796 | |||
BC016968.1 | - | 11 | 1817 | |||
BC021924.1 | - | 7 | 905 | |||
BC110435.1 | - | 12 | 1789 | 5518312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
