siRNA Details
Gene Symbol: R3HCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136108.2 | NP_001129580.2 | 8 | 1576 | ||
NM_001301650.1 | NP_001288579.1 | 9 | 1450 | |||
NR_125897.1 | 9 | 1461 | ||||
GenBank mRNA | AK293465.1 | - | 8 | 1560 | ||
BC032421.1 | - | 8 | 1543 | |||
BC050572.1 | - | 7 | 1515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
