siRNA Details
Gene Symbol: R3HCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136108.2 | NP_001129580.2 | 3 | 242 | ||
NM_001301650.1 | NP_001288579.1 | 3 | 242 | |||
NR_125897.1 | 3 | 242 | ||||
GenBank mRNA | AK293465.1 | - | 3 | 226 | ||
AL050297.1 | - | 3 | 220 | |||
BC032421.1 | - | 3 | 209 | |||
BC050572.1 | - | 2 | 181 | |||
BC143822.1 | - | 3 | 426 | 9052337 | ||
BC143827.1 | - | 3 | 426 | |||
BC143829.1 | - | 3 | 426 | 9052344 | ||
BX426634.2 | - | 3 | 255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
