siRNA Details
Gene Symbol: SNX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301089.1 | NP_001288018.1 | 2 | 336 | ||
NM_014758.2 | NP_055573.2 | 2 | 2350 | |||
XM_005271546.4 | XP_005271603.1 | 2 | 2349 | |||
XM_011542819.2 | XP_011541121.1 | 2 | 2349 | |||
XM_011542820.2 | XP_011541122.1 | 2 | 2349 | |||
XM_011542821.2 | XP_011541123.1 | 2 | 2350 | |||
XM_011542822.2 | XP_011541124.1 | 2 | 2349 | |||
XM_011542824.2 | XP_011541126.1 | 2 | 1254 | |||
XM_011542825.2 | XP_011541127.1 | 2 | 334 | |||
XM_011542826.2 | XP_011541128.1 | 2 | 333 | |||
XM_011542827.2 | XP_011541129.1 | 2 | 334 | |||
XM_017017725.1 | XP_016873214.1 | 2 | 2325 | |||
XM_017017726.1 | XP_016873215.1 | 2 | 331 | |||
GenBank mRNA | AF395843.1 | - | 2 | 1802 | ||
AK292457.1 | - | 2 | 2372 | |||
AK304809.1 | - | 3 | 356 | |||
AK309202.1 | - | 2 | 300 | |||
AK316414.1 | - | 2 | 300 | |||
AK316505.1 | - | 2 | 293 | |||
BC031620.1 | - | 2 | 2312 | 5172852 | ||
BU431333.1 | - | 1 | 140 | |||
D87443.1 | - | 2 | 2330 | |||
DC400642.1 | - | 2 | 336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
