siRNA Details
Gene Symbol: GAB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037814.1 | NP_001032903.1 | 2 | 458 | ||
XM_011546114.2 | XP_011544416.1 | 2 | 510 | |||
XM_011546115.1 | XP_011544417.1 | 2 | 243 | |||
XM_011546116.2 | XP_011544418.1 | 2 | 517 | |||
XM_017028575.1 | XP_016884064.1 | 2 | 510 | |||
GenBank mRNA | AB076978.1 | - | 2 | 458 | ||
AK057252.1 | - | 2 | 235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
