siRNA Details
Gene Symbol: GAB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037814.1 | NP_001032903.1 | 6 | 1320 | ||
XM_011546114.2 | XP_011544416.1 | 4 | 913 | |||
XM_011546115.1 | XP_011544417.1 | 5 | 854 | |||
XM_011546117.1 | XP_011544419.1 | 4 | 610 | |||
XM_017028575.1 | XP_016884064.1 | 6 | 1372 | |||
XM_017028576.1 | XP_016884065.1 | 4 | 541 | |||
GenBank mRNA | AB076978.1 | - | 6 | 1320 | ||
AK057252.1 | - | 5 | 1912 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
