siRNA Details
Gene Symbol: TTC28-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026963.1 | 5 | 2181 | |||
GenBank mRNA | AB051435.1 | - | 4 | 1899 | ||
AL832597.1 | - | 5 | 2137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
