siRNA Details
Gene Symbol: KIAA1644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099294.1 | NP_001092764.1 | 4 | 445 | ||
XM_005261790.3 | XP_005261847.1 | 4 | 543 | |||
GenBank mRNA | AB051431.1 | - | 2 | 111 | ||
BC104183.1 | - | 3 | 312 | 40030810 | ||
BC104184.1 | - | 3 | 312 | 40030814 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
