siRNA Details
Gene Symbol: RAPGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098531.2 | NP_001092001.1 | 28 | 5478 | ||
NM_001098532.2 | NP_001092002.1 | 27 | 5242 | |||
NM_006105.5 | NP_006096.2 | 28 | 5012 | |||
GenBank mRNA | U79275.1 | - | 1 | 299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
