siRNA Details
Gene Symbol: PTPRQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145026.1 | NP_001138498.1 | 42 | 6584 | ||
XM_017019273.1 | XP_016874762.1 | 47 | 7431 | |||
XM_017019274.1 | XP_016874763.1 | 46 | 7387 | |||
GenBank mRNA | AF169351.1 | - | 3 | 287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
