siRNA Details
Gene Symbol: TMEM132B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286219.1 | NP_001273148.1 | 5 | 1633 | ||
NM_052907.3 | NP_443139.2 | 9 | 2886 | |||
XM_011537852.2 | XP_011536154.1 | 9 | 2943 | |||
XM_011537854.2 | XP_011536156.1 | 5 | 7616 | |||
XM_017018767.1 | XP_016874256.1 | 10 | 3052 | |||
GenBank mRNA | AB067493.1 | - | 4 | 1423 | ||
BC113366.1 | - | 5 | 1510 | 8322418 | ||
BC113368.1 | - | 5 | 1510 | 8322420 | ||
BC131621.1 | - | 5 | 1550 | 40117411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
