siRNA Details
Gene Symbol: ANKRD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173595.3 | NP_775866.2 | 26 | 2962 | ||
XM_011538197.2 | XP_011536499.1 | 25 | 3064 | |||
XM_017019183.1 | XP_016874672.1 | 25 | 3145 | |||
XM_017019184.1 | XP_016874673.1 | 26 | 2994 | |||
GenBank mRNA | AB190990.1 | - | 26 | 2979 | ||
AK125379.1 | - | 26 | 2979 | |||
AK126893.1 | - | 19 | 2121 | |||
BC042070.1 | - | 6 | 588 |

More Information
Other Species Targeted

Gene Ontology Categories:
