siRNA Details
Gene Symbol: SMIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024058.2 | NP_076963.1 | 2 | 253 | ||
GenBank mRNA | BC000868.1 | - | 2 | 253 | 3460233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
