siRNA Details
Gene Symbol: SETD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015048.1 | NP_055863.1 | 17 | 7648 | ||
XM_005253858.4 | XP_005253915.1 | 17 | 7936 | |||
XM_006719296.3 | XP_006719359.1 | 17 | 7852 | |||
GenBank mRNA | AB028999.1 | - | 7 | 4287 | ||
AK000810.1 | - | 1 | 35 | |||
BC047909.1 | - | 1 | 1568 | |||
BC080192.1 | - | 1 | 1815 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
