siRNA Details
Gene Symbol: METTL21C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010977.2 | NP_001010977.1 | 2 | 203 | ||
XM_017020407.1 | XP_016875896.1 | 3 | 1097 | |||
XM_017020408.1 | XP_016875897.1 | 3 | 1650 | |||
GenBank mRNA | BC132748.1 | - | 2 | 203 | 40146770 | |
BC132750.1 | - | 2 | 203 | 40146772 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
