siRNA Details
Gene Symbol: TTC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310135.1 | NP_001297064.1 | 21 | 4759 | ||
XM_011537430.2 | XP_011535732.1 | 16 | 2567 | |||
XM_011537431.2 | XP_011535733.1 | 14 | 2224 | |||
XM_017021254.1 | XP_016876743.1 | 21 | 4759 | |||
XM_017021255.1 | XP_016876744.1 | 21 | 4759 | |||
XM_017021256.1 | XP_016876745.1 | 19 | 4849 | |||
GenBank mRNA | BC103914.2 | - | 2 | 108 | 40006180 | |
BC103915.1 | - | 2 | 108 | 40006182 | ||
BC103916.2 | - | 2 | 108 | 40006183 | ||
BX161415.1 | - | 3 | 334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
