siRNA Details
Gene Symbol: TTC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310135.1 | NP_001297064.1 | 25, 26 | 5370 | ||
XM_011537430.2 | XP_011535732.1 | 21, 22 | 3202 | |||
XM_011537431.2 | XP_011535733.1 | 19, 20 | 2859 | |||
XM_017021254.1 | XP_016876743.1 | 24, 25 | 5220 | |||
XM_017021255.1 | XP_016876744.1 | 23, 24 | 5085 | |||
XM_017021256.1 | XP_016876745.1 | 24, 25 | 5484 | |||
GenBank mRNA | BC103914.2 | - | 6, 7 | 719 | 40006180 | |
BC103915.1 | - | 6, 7 | 719 | 40006182 | ||
BC103916.2 | - | 6, 7 | 719 | 40006183 | ||
BX161415.1 | - | 5, 6 | 654 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
