siRNA Details
Gene Symbol: TTC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310135.1 | NP_001297064.1 | 30 | 5938 | ||
XM_011537430.2 | XP_011535732.1 | 26 | 3770 | |||
XM_011537431.2 | XP_011535733.1 | 24 | 3427 | |||
XM_017021254.1 | XP_016876743.1 | 29 | 5788 | |||
XM_017021255.1 | XP_016876744.1 | 28 | 5653 | |||
XM_017021256.1 | XP_016876745.1 | 29 | 6052 | |||
GenBank mRNA | BC014342.2 | - | 3 | 345 | ||
BC103914.2 | - | 11 | 1287 | 40006180 | ||
BC103915.1 | - | 11 | 1287 | 40006182 | ||
BC103916.2 | - | 11 | 1287 | 40006183 | ||
BX161415.1 | - | 10 | 1222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
