siRNA Details
Gene Symbol: PLEKHG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308147.1 | NP_001295076.1 | 16 | 2165 | ||
XM_011536624.2 | XP_011534926.1 | 18 | 2446 | |||
XM_011536625.2 | XP_011534927.1 | 18 | 2319 | |||
XM_011536627.2 | XP_011534929.1 | 18 | 2711 | |||
XM_011536628.2 | XP_011534930.1 | 18 | 2897 | |||
XM_011536629.2 | XP_011534931.1 | 17 | 2035 | |||
XM_017021154.1 | XP_016876643.1 | 17 | 2312 | |||
XM_017021155.1 | XP_016876644.1 | 18 | 2401 | |||
XM_017021156.1 | XP_016876645.1 | 18 | 2469 | |||
XM_017021157.1 | XP_016876646.1 | 16 | 2265 | |||
XM_017021158.1 | XP_016876647.1 | 16 | 1904 | |||
GenBank mRNA | AB011171.1 | - | 1 | 263 | ||
AK074169.1 | - | 10 | 1936 | |||
BC004298.1 | - | 3 | 504 | |||
BC011891.2 | - | 1 | 255 | |||
BC063554.1 | - | 2 | 346 | |||
BC073907.1 | - | 3 | 453 | |||
BC129952.1 | - | 10 | 1657 | 40126555 | ||
BC129953.1 | - | 16 | 1976 | 40126558 | ||
BX248779.1 | - | 14 | 1969 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
