siRNA Details
Gene Symbol: FOXN3-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024620.1 | 1 | 763 | |||
GenBank mRNA | AF118074.1 | - | 1 | 763 | ||
BC069658.1 | - | 1 | 708 | |||
BC069757.1 | - | 1 | 712 | |||
BC069775.1 | - | 1 | 712 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
