siRNA Details
Gene Symbol: PIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286496.1 | NP_001273425.1 | 13 | 2500 | ||
NM_001286497.1 | NP_001273426.1 | 13 | 2067 | |||
NM_001286499.1 | NP_001273428.1 | 13 | 2500 | |||
NM_025049.3 | NP_079325.2 | 13 | 2476 | |||
XM_011522083.2 | XP_011520385.1 | 13 | 2592 | |||
XM_011522084.2 | XP_011520386.1 | 13 | 2424 | |||
GenBank mRNA | AB185926.1 | - | 13 | 2476 | ||
AB185927.1 | - | 13 | 2067 | |||
AF108138.1 | - | 9 | 1580 | |||
AY498716.1 | - | 12 | 2400 | |||
BC033254.1 | - | 7 | 1341 | |||
EU084033.1 | - | 13 | 2471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
