siRNA Details
Gene Symbol: PIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286496.1 | NP_001273425.1 | 13 | 2198 | ||
NM_001286499.1 | NP_001273428.1 | 13 | 2198 | |||
NM_025049.3 | NP_079325.2 | 13 | 2174 | |||
XM_011522083.2 | XP_011520385.1 | 13 | 2290 | |||
XM_011522084.2 | XP_011520386.1 | 13 | 2122 | |||
GenBank mRNA | AB185926.1 | - | 13 | 2174 | ||
AF108138.1 | - | 9 | 1278 | |||
AY498716.1 | - | 12 | 2098 | |||
BC033254.1 | - | 7 | 1039 | |||
BC137503.1 | - | 13 | 2112 | 9021503 | ||
BC137504.1 | - | 13 | 2112 | 9021504 | ||
EU084033.1 | - | 13 | 2169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
