siRNA Details
Gene Symbol: CDR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001802.1 | NP_001793.1 | 5 | 2080 | ||
GenBank mRNA | BC017503.1 | - | 5 | 2062 | 4873337 | |
D12981.1 | - | 5 | 1755 | |||
M63256.1 | - | 5 | 1934 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
