siRNA Details
Gene Symbol: NUDT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105663.2 | NP_001099133.1 | 2 | 195 | ||
NM_001243657.1 | NP_001230586.1 | 2 | 195 | |||
NM_001243660.1 | NP_001230589.1 | 2 | 195 | |||
NM_001243661.1 | NP_001230590.1 | 2 | 195 | |||
XM_011523032.2 | XP_011521334.1 | 2 | 199 | |||
XM_011523033.2 | XP_011521335.1 | 2 | 199 | |||
XM_017023170.1 | XP_016878659.1 | 2 | 199 | |||
GenBank mRNA | AA227330.1 | - | 2 | 147 | ||
AK296963.1 | - | 2 | 168 | |||
BU579534.1 | - | 2 | 141 | |||
BX387030.2 | - | 2 | 195 | |||
BX494638.1 | - | 2 | 174 | |||
CN297379.1 | - | 2 | 146 | |||
EU981826.1 | - | 2 | 104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
