siRNA Details
Gene Symbol: NUDT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105663.2 | NP_001099133.1 | 4 | 576 | ||
NM_001243657.1 | NP_001230586.1 | 5 | 670 | |||
NM_001243660.1 | NP_001230589.1 | 4 | 531 | |||
NM_001243661.1 | NP_001230590.1 | 3 | 417 | |||
XM_011523032.2 | XP_011521334.1 | 5 | 694 | |||
XM_011523033.2 | XP_011521335.1 | 6 | 734 | |||
XM_017023170.1 | XP_016878659.1 | 6 | 788 | |||
GenBank mRNA | AK296963.1 | - | 3 | 390 | ||
BU579534.1 | - | 4 | 522 | |||
BX387030.2 | - | 4 | 531 | |||
CN297379.1 | - | 5 | 621 | |||
ES309123.1 | - | 2 | 311 | |||
EU981826.1 | - | 3 | 326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
