siRNA Details
Gene Symbol: CDRT15P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003261.1 | 1 | 447 | |||
GenBank mRNA | AA453375.1 | - | 2 | 307 | ||
BX118788.1 | - | 2 | 307 |
Gene Symbol: CDRT15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007530.1 | NP_001007531.1 | 2 | 290 | ||
GenBank mRNA | BC146859.1 | - | 2 | 316 | 9056787 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
