siRNA Details
Gene Symbol: TAF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293725.1 | NP_001280654.1 | 8 | 1768 | ||
NM_005640.2 | NP_005631.1 | 8 | 1753 | |||
NR_121653.1 | 8 | 1753 | ||||
XM_005258339.3 | XP_005258396.1 | 8 | 2145 | |||
XM_011526153.2 | XP_011524455.1 | 8 | 2145 | |||
XM_017025932.1 | XP_016881421.1 | 8 | 2145 | |||
XM_017025933.1 | XP_016881422.1 | 8 | 1377 | |||
GenBank mRNA | AB234096.1 | - | 8 | 2165 | ||
AK302593.1 | - | 7 | 1242 | |||
AY888048.1 | - | 8 | 1753 | |||
AY888049.1 | - | 8 | 1753 | |||
Y09321.1 | - | 8 | 1519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
