siRNA Details
Gene Symbol: ASXL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030632.2 | NP_085135.1 | 11 | 2684 | ||
XM_005258356.1 | XP_005258413.1 | 12 | 2657 | |||
XM_011526205.2 | XP_011524507.1 | 12 | 9447 | |||
XM_011526206.2 | XP_011524508.1 | 12 | 6212 | |||
XM_011526209.1 | XP_011524511.1 | 13 | 2841 | |||
XM_011526210.1 | XP_011524512.1 | 13 | 2898 | |||
XM_011526212.1 | XP_011524514.1 | 12 | 2802 | |||
XM_011526213.1 | XP_011524515.1 | 12 | 2895 | |||
XM_017026012.1 | XP_016881501.1 | 12 | 6059 | |||
XM_017026013.1 | XP_016881502.1 | 13 | 6421 | |||
XM_017026014.1 | XP_016881503.1 | 13 | 2858 | |||
GenBank mRNA | AB051500.2 | - | 1 | 843 | ||
AK131454.1 | - | 13 | 2727 | |||
AL133050.1 | - | 1 | 179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
