siRNA Details
Gene Symbol: ASXL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030632.2 | NP_085135.1 | 11 | 2751 | ||
XM_005258356.1 | XP_005258413.1 | 12 | 2724 | |||
XM_011526205.2 | XP_011524507.1 | 12 | 9514 | |||
XM_011526206.2 | XP_011524508.1 | 12 | 6279 | |||
XM_011526209.1 | XP_011524511.1 | 13 | 2908 | |||
XM_011526210.1 | XP_011524512.1 | 13 | 2965 | |||
XM_011526212.1 | XP_011524514.1 | 12 | 2869 | |||
XM_011526213.1 | XP_011524515.1 | 12 | 2962 | |||
XM_017026012.1 | XP_016881501.1 | 12 | 6126 | |||
XM_017026013.1 | XP_016881502.1 | 13 | 6488 | |||
XM_017026014.1 | XP_016881503.1 | 13 | 2925 | |||
GenBank mRNA | AB051500.2 | - | 1 | 910 | ||
AK131454.1 | - | 13 | 2794 | |||
AL133050.1 | - | 1 | 246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
