siRNA Details
Gene Symbol: CCDC40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243342.1 | NP_001230271.1 | 8 | 1283 | ||
NM_017950.3 | NP_060420.2 | 8 | 1283 | |||
XM_005257492.4 | XP_005257549.1 | 8 | 1283 | |||
XM_011524963.2 | XP_011523265.1 | 8 | 1320 | |||
XM_011524964.2 | XP_011523266.1 | 2 | 252 | |||
XM_011524965.2 | XP_011523267.1 | 8 | 1283 | |||
XM_017024807.1 | XP_016880296.1 | 8 | 1283 | |||
GenBank mRNA | AB046860.1 | - | 4 | 789 | ||
AK000760.1 | - | 8 | 1263 | |||
AK130684.1 | - | 2 | 1352 | |||
BC035251.1 | - | 8 | 1276 | 4838111 | ||
BC058288.1 | - | 8 | 1261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
