siRNA Details
Gene Symbol: CEP131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009811.3 | NP_001009811.2 | 16 | 2171 | ||
NM_001319228.1 | NP_001306157.1 | 16 | 2180 | |||
NM_001319229.1 | NP_001306158.1 | 16 | 2180 | |||
NM_014984.3 | NP_055799.2 | 16 | 2171 | |||
XM_011524547.2 | XP_011522849.1 | 16 | 1977 | |||
XM_011524548.2 | XP_011522850.1 | 16 | 2171 | |||
XM_011524550.2 | XP_011522852.1 | 16 | 2180 | |||
XM_011524551.2 | XP_011522853.1 | 16 | 2180 | |||
XM_011524552.2 | XP_011522854.1 | 16 | 2180 | |||
XM_011524553.2 | XP_011522855.1 | 16 | 2180 | |||
XM_011524555.2 | XP_011522857.1 | 16 | 2180 | |||
XM_017024369.1 | XP_016879858.1 | 16 | 2180 | |||
GenBank mRNA | AB029041.1 | - | 16 | 2190 | ||
BC011615.2 | - | 7 | 912 | |||
BC136578.1 | - | 16 | 2112 | 9020568 | ||
BC136580.1 | - | 16 | 2112 | 9020570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
