siRNA Details
Gene Symbol: RNF165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256758.1 | NP_001243687.1 | 6 | 1010 | ||
NM_152470.2 | NP_689683.2 | 8 | 1434 | |||
XM_011526016.2 | XP_011524318.1 | 7 | 1338 | |||
GenBank mRNA | AK122819.1 | - | 6 | 1010 | ||
AK123883.1 | - | 2 | 520 | |||
AK127467.1 | - | 8 | 1434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
