siRNA Details
Gene Symbol: MISP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291291.1 | NP_001278220.1 | 3 | 957 | ||
NR_111941.1 | 3 | 1022 | ||||
GenBank mRNA | BC011002.2 | - | 3 | 957 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
