siRNA Details
Gene Symbol: ZFP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297619.1 | NP_001284548.1 | 5 | 379 | ||
NM_020917.2 | NP_065968.1 | 5 | 376 | |||
XM_017027038.1 | XP_016882527.1 | 5 | 656 | |||
GenBank mRNA | AB046779.1 | - | 4 | 349 | ||
AK293103.1 | - | 5 | 372 | |||
AL699477.1 | - | 5 | 377 | |||
BC144691.1 | - | 4 | 349 | |||
BC152446.1 | - | 4 | 349 | 8862575 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
