siRNA Details
Gene Symbol: URB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014825.2 | NP_055640.2 | 35 | 5652 | ||
GenBank mRNA | AB011111.2 | - | 30 | 4886 | ||
AF231919.1 | - | 35 | 5490 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
