siRNA Details
Gene Symbol: DIP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146116.1 | NP_001139588.1 | 23 | 2898 | ||
NM_015151.3 | NP_055966.2 | 23 | 2910 | |||
XM_011529492.2 | XP_011527794.1 | 24 | 3025 | |||
XM_011529501.1 | XP_011527803.1 | 24 | 3025 | |||
XM_017028291.1 | XP_016883780.1 | 24 | 3025 | |||
XM_017028292.1 | XP_016883781.1 | 24 | 3025 | |||
XM_017028293.1 | XP_016883782.1 | 24 | 3022 | |||
XM_017028294.1 | XP_016883783.1 | 23 | 2904 | |||
XM_017028295.1 | XP_016883784.1 | 23 | 2901 | |||
XM_017028296.1 | XP_016883785.1 | 23 | 2901 | |||
XM_017028297.1 | XP_016883786.1 | 23 | 2893 | |||
XM_017028298.1 | XP_016883787.1 | 22 | 2772 | |||
XM_017028299.1 | XP_016883788.1 | 23 | 2770 | |||
XM_017028300.1 | XP_016883789.1 | 24 | 3025 | |||
XM_017028301.1 | XP_016883790.1 | 24 | 3025 | |||
XM_017028302.1 | XP_016883791.1 | 22 | 2665 | |||
XM_017028303.1 | XP_016883792.1 | 24 | 3025 | |||
XM_017028304.1 | XP_016883793.1 | 24 | 3025 | |||
XM_017028305.1 | XP_016883794.1 | 24 | 3021 | |||
GenBank mRNA | AB273729.1 | - | 23 | 2748 | ||
AF490768.1 | - | 23 | 2803 | |||
AK303351.1 | - | 21 | 2725 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
