siRNA Details
Gene Symbol: LMTK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080434.1 | NP_001073903.1 | 6 | 548 | ||
XM_011526411.2 | XP_011524713.1 | 6 | 879 | |||
XM_011526412.2 | XP_011524714.1 | 6 | 846 | |||
GenBank mRNA | AB067470.1 | - | 6 | 521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
